Skip to Content
AI
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
L-2-Aminobutyric acid
Synonym(s):
L-α-Aminobutyric acid
Empirical Formula (Hill Notation):
C
4
H
9
NO
2
CAS Number:
1492-24-6
Molecular Weight:
103.12
EC Number:
216-083-3
Products
Product Category
amino acids (1)
media supplements (1)
Brand
Sigma-Aldrich (2)
Biological Source
Color
Formula Weight
Physical Form
Product Line
Purity
Tissue
Quality Segment
Application
Optical Isomer
Technique
Filter Products
Showing 1-2 of 2
Compare
Product Number
Description
Pricing
A1879
L-2-Aminobutyric acid,
≥99% (titration), suitable for ligand binding assays
Expand
View Pricing
A2536
L-2-Aminobutyric acid,
BioReagent, suitable for cell culture
Expand
View Pricing