Skip to Content
AI
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Kontes
®
NMR tubes 5 mm diam., thrift N-51A glass
Products
Product Category
tubes (10)
Diameter (mm)
Length (in)
Color
Frequency (MHz)
Filter Products
Showing 1-10 of 10
Compare
Product Number
Description
Pricing
KN8971400007
Kontes
®
NMR tubes 5 mm diam., thrift N-51A glass,
frequency 400 MHz, L × camber × concentricity 7 in. × 6 μm × 13 μm
Expand
View Pricing
KN8971100007
Kontes
®
NMR tubes 5 mm diam., thrift N-51A glass,
frequency >200 MHz, size 7 in., size × camber × concentricity × 25 μm × 51 μm
Expand
View Pricing
KN8971400008
Kontes
®
NMR tubes 5 mm diam., thrift N-51A glass,
frequency 400 MHz, L × camber × concentricity 8 in. × 9 μm × 13 μm
Expand
View Pricing
KN8971200008
Kontes
®
NMR tubes 5 mm diam., thrift N-51A glass,
frequency 200 MHz, L × camber × concentricity 8 in. × 15 μm × 51 μm
Expand
View Pricing
KN8971300008
Kontes
®
NMR tubes 5 mm diam., thrift N-51A glass,
frequency 300 MHz, L × camber × concentricity 8 in. × 28 μm × 38 μm
Expand
View Pricing
KN8971200007
Kontes
®
NMR tubes 5 mm diam., thrift N-51A glass,
frequency 200 MHz, L × camber × concentricity 7 in. × 13 μm × 51 μm
Expand
View Pricing
KN8971100008
Kontes
®
NMR tubes 5 mm diam., thrift N-51A glass,
frequency >200 MHz, L × camber × concentricity 8 in. × 28 μm × 51 μm
Expand
View Pricing
KN8971500007
Kontes
®
NMR tubes 5 mm diam., thrift N-51A glass,
frequency 500 MHz, L × camber × concentricity 7 in. × 6 μm × 13 μm
Expand
View Pricing
KN8971500008
Kontes
®
NMR tubes 5 mm diam., thrift N-51A glass,
frequency 500 MHz, L × camber × concentricity 8 in. × 9 μm × 13 μm
Expand
View Pricing
KN8971300007
Kontes
®
NMR tubes 5 mm diam., thrift N-51A glass,
frequency 300 MHz, L × camber × camber 7 in. × 25 μm × 38 μm
Expand
View Pricing