Isoproturon
Synonym(s):
3-(4-Isopropylphenyl)-1,1-dimethylurea
Empirical Formula (Hill Notation):
C12H18N2O
Products
Product Category
Formula Weight
Product Line
Format
Quality Segment
Application
Reference Material Type
Special Grade
Technique
Showing 1-1 of 1
Compare
Product Number
Description
Pricing