Skip to Content
AI
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Iron(II) sulfide
Synonym(s):
Ferrous sulfide
Empirical Formula (Hill Notation):
FeS
CAS Number:
1317-37-9
Molecular Weight:
87.91
EC Number:
215-268-6
Products
Product Category
salts (3)
Brand
Sigma-Aldrich (4)
Formula Weight
Physical Form
Purity
Quality Segment
Reagent Type
Core
Special Grade
Technical (1)
Filter Products
Showing 1-4 of 4
Compare
Product Number
Description
Pricing
343161
Iron(II) sulfide,
−100 mesh, 99.9% trace metals basis
Expand
View Pricing
268704
Iron(II) sulfide,
technical grade
Expand
View Pricing
12363
Iron(II) sulfide,
sticks (thin)
Expand
View Pricing
15-1520
Iron(II) sulfide,
SAJ first grade, ≥50.0%
Expand
View Pricing