Skip to Content
AI
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Invar
®
- Controlled Expansion Alloy, Fe64Ni36
Products
Manufacturer
Physical Form
Diameter (mm)
Length (m)
Filter Products
Showing 1-20 of 198
Page 1 of 10
Page 1 of 10
Compare
Product Number
Description
Pricing
GF38432639
Invar
®
- Controlled Expansion Alloy, Fe64Ni36,
sheet, 150 x 150mm, 2.5mm thickness, annealed
Expand
View Pricing
GF46542618
Invar
®
- Controlled Expansion Alloy, Fe64Ni36,
foil, 100 x 100mm, 0.02mm thickness, light tight (lt)
Expand
View Pricing
GF82950622
Invar
®
- Controlled Expansion Alloy, Fe64Ni36,
20m wire, 0.25mm diameter, annealed
Expand
View Pricing
GF63908309
Invar
®
- Controlled Expansion Alloy, Fe64Ni36,
200mm rod, 19mm diameter
Expand
View Pricing
GF50265255
Invar
®
- Controlled Expansion Alloy, Fe64Ni36,
50m wire, 0.6mm diameter, annealed
Expand
View Pricing
GF81269006
Invar
®
- Controlled Expansion Alloy, Fe64Ni36,
100mm rod, 3.2mm diameter
Expand
View Pricing
GF92166598
Invar
®
- Controlled Expansion Alloy, Fe64Ni36,
powder, 45micron max. particle size, atomized, 50g
Expand
View Pricing
GF27132858
Invar
®
- Controlled Expansion Alloy, Fe64Ni36,
foil, 25 x 25mm, 0.08mm thickness, annealed, light tight (lt)
Expand
View Pricing
GF30768093
Invar
®
- Controlled Expansion Alloy, Fe64Ni36,
foil, 100 x 100mm, 0.025mm thickness, light tight (lt)
Expand
View Pricing
GF15627225
Invar
®
- Controlled Expansion Alloy, Fe64Ni36,
foil, 50 x 50mm, 0.5mm thickness, annealed
Expand
View Pricing
GF71755533
Invar
®
- Controlled Expansion Alloy, Fe64Ni36,
foil, 40 x 100mm, 0.25mm thickness, annealed, flattened
Expand
View Pricing
GF22561185
Invar
®
- Controlled Expansion Alloy, Fe64Ni36,
100mm rod, 1.6mm diameter
Expand
View Pricing
GF04113324
Invar
®
- Controlled Expansion Alloy, Fe64Ni36,
100mm tube, 2.2mm outside diameter, 0.16mm wall thickness, 1.88mm inside diameter
Expand
View Pricing
GF90254077
Invar
®
- Controlled Expansion Alloy, Fe64Ni36,
foil, 100 x 100mm, 0.125mm thickness, half hard
Expand
View Pricing
GF05826719
Invar
®
- Controlled Expansion Alloy, Fe64Ni36,
foil, 50 x 50mm, 0.125mm thickness, half hard
Expand
View Pricing
GF11220983
Invar
®
- Controlled Expansion Alloy, Fe64Ni36,
foil, 25 x 25mm, 0.05mm thickness, annealed, light tight (lt)
Expand
View Pricing
GF44902289
Invar
®
- Controlled Expansion Alloy, Fe64Ni36,
foil, 25 x 25mm, 0.025mm thickness, light tight (lt)
Expand
View Pricing
GF86487902
Invar
®
- Controlled Expansion Alloy, Fe64Ni36,
foil, 25mm disks, 0.0125mm thickness
Expand
View Pricing
GF66891785
Invar
®
- Controlled Expansion Alloy, Fe64Ni36,
foil, 50 x 50mm, 0.0125mm thickness, light tight (lt)
Expand
View Pricing
GF21714397
Invar
®
- Controlled Expansion Alloy, Fe64Ni36,
foil, 25 x 25mm, 0.0125mm thickness, light tight (lt)
Expand
View Pricing
Page 1 of 10
Page 1 of 10