Skip to Content
AI
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Indene
Synonym(s):
Indene
Empirical Formula (Hill Notation):
C
9
H
8
CAS Number:
95-13-6
Molecular Weight:
116.16
EC Number:
202-393-6
Products
Product Category
building blocks (3)
Brand
Sigma-Aldrich (4)
Boiling Point (°C)
Formula Weight
Melting Point (°C)
Physical Form
Purity
Quality Segment
Markush Class
Markush Group
Special Grade
Technical (1)
Filter Products
Showing 1-4 of 4
Compare
Product Number
Description
Pricing
193828
Indene,
≥99%
Expand
View Pricing
168769
Indene,
98%
Expand
View Pricing
8.20701
Indene,
for synthesis
Expand
View Pricing
I2800
Indene,
contains 50-100 ppm
tert
-butylcatechol as stabilizer, technical grade, ≥90%
Expand
View Pricing