
Hyoscyamine Related Compound A

Synonym(s):
Norhyoscyamine sulfate
Empirical Formula (Hill Notation):
C32H44N2O10S
Molecular Weight:
648.76
Products
Formula Weight
Manufacturer
Agency
Format
Quality Segment
Application
API Family
Reference Material Type
Special Grade
Showing 1-2 of 2