
H-Lys(Boc)-OH

Synonym(s):
Nε-Boc-L-lysine
Linear Formula:
(CH3)3COCONH(CH2)4CH(NH2)COOH
CAS Number:
Molecular Weight:
246.30
Products
Brand
Formula Weight
Melting Point (°C)
Physical Form
Purity
Quality Segment
Application
Markush Class
Markush Group
Optical Isomer
Amino Acid
Amino Acid Type
Reaction Type
Showing 1-1 of 1
Compare
Product Number
Description
Pricing