Skip to Content
AI
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Hexamethylphosphoramide
Synonym(s):
HMPA, Hexamethylphosphoric acid triamide, Tris(dimethylamino)phosphine oxide
Linear Formula:
[(CH
3
)
2
N]
3
PO
CAS Number:
680-31-9
Molecular Weight:
179.20
EC Number:
211-653-8
Products
Brand
Sigma-Aldrich (3)
Boiling Point (°C)
Formula Weight
Melting Point (°C)
Physical Form
Purity
Quality Segment
Markush Class
Markush Group
Special Grade
Absolute (1)
Purum (1)
Filter Products
Showing 1-3 of 3
Compare
Product Number
Description
Pricing
H11602
Hexamethylphosphoramide,
99%
Expand
View Pricing
779903
Hexamethylphosphoramide,
absolute, over molecular sieve (H
2
O ≤0.03%), ≥98.0% (GC)
Expand
View Pricing
52730
Hexamethylphosphoramide,
purum, ≥98.0% (GC)
Expand
View Pricing