Skip to Content
AI
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Hexamethyldisiloxane
Synonym(s):
HMDSO, Hexamethyldisiloxane
Linear Formula:
(CH
3
)
3
SiOSi(CH
3
)
3
CAS Number:
107-46-0
Molecular Weight:
162.38
EC Number:
203-492-7
Products
Brand
Sigma-Aldrich (5)
Boiling Point (°C)
Formula Weight
Melting Point (°C)
Physical Form
Purity
Quality Segment
Special Grade
Puriss (1)
Technique
Filter Products
Showing 1-5 of 5
Compare
Product Number
Description
Pricing
205389
Hexamethyldisiloxane,
≥98%
Expand
View Pricing
469300
Hexamethyldisiloxane,
viscosity 0.65 cSt (25 °C)
Expand
View Pricing
326739
Hexamethyldisiloxane,
NMR grade, ≥99.5%
Expand
View Pricing
8.14051
Hexamethyldisiloxane,
for synthesis
Expand
View Pricing
52630
Hexamethyldisiloxane,
puriss., ≥98.5% (GC)
Expand
View Pricing