Skip to Content
AI
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Water Purification
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
Get Site Smart
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Hexaethylene glycol monododecyl ether
Synonym(s):
C
12
E
6
, Dodecylhexaglycol, Polyoxyethylene (6) lauryl ether
Linear Formula:
CH
3
(CH
2
)
11
(OCH
2
CH
2
)
6
OH
CAS Number:
3055-96-7
Molecular Weight:
450.65
EC Number:
221-282-3
Products
Product Category
detergents (1)
Formula Weight
Product Line
Purity
Filter Products
Showing 1-1 of 1
Compare
Product Number
Description
Pricing
52044
Hexaethylene glycol monododecyl ether,
BioXtra, ≥98.0% (TLC)
Expand
View Pricing