Skip to Content
AI
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Hastelloy
®
B-3 alloy, Ni65Mo28.5Cr1.5Fe1.5(Co+Mn+W)3.5max
Products
Manufacturer
Physical Form
Diameter (mm)
Length (m)
Length (mm)
Filter Products
Showing 1-9 of 9
Compare
Product Number
Description
Pricing
GF56744231
Hastelloy
®
B-3 alloy, Ni65Mo28.5Cr1.5Fe1.5(Co+Mn+W)3.5max,
200mm rod, 5.5mm diameter, annealed
Expand
View Pricing
GF75073103
Hastelloy
®
B-3 alloy, Ni65Mo28.5Cr1.5Fe1.5(Co+Mn+W)3.5max,
500mm rod, 5.5mm diameter, annealed
Expand
View Pricing
GF37313322
Hastelloy
®
B-3 alloy, Ni65Mo28.5Cr1.5Fe1.5(Co+Mn+W)3.5max,
100mm rod, 5.5mm diameter, annealed
Expand
View Pricing
GF29720628
Hastelloy
®
B-3 alloy, Ni65Mo28.5Cr1.5Fe1.5(Co+Mn+W)3.5max,
1m wire, 0.5mm diameter, annealed
Expand
View Pricing
GF56619625
Hastelloy
®
B-3 alloy, Ni65Mo28.5Cr1.5Fe1.5(Co+Mn+W)3.5max,
50m wire, 0.5mm diameter, annealed
Expand
View Pricing
GF26756860
Hastelloy
®
B-3 alloy, Ni65Mo28.5Cr1.5Fe1.5(Co+Mn+W)3.5max,
20m wire, 0.5mm diameter, annealed
Expand
View Pricing
GF64663386
Hastelloy
®
B-3 alloy, Ni65Mo28.5Cr1.5Fe1.5(Co+Mn+W)3.5max,
5m wire, 0.5mm diameter, annealed
Expand
View Pricing
GF37027717
Hastelloy
®
B-3 alloy, Ni65Mo28.5Cr1.5Fe1.5(Co+Mn+W)3.5max,
1000mm rod, 5.5mm diameter, annealed
Expand
View Pricing
GF29270448
Hastelloy
®
B-3 alloy, Ni65Mo28.5Cr1.5Fe1.5(Co+Mn+W)3.5max,
10m wire, 0.5mm diameter, annealed
Expand
View Pricing