Skip to Content
AI
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Gold Nanospheres
Synonym(s):
Gold Nanospheres
Products
Brand
Sigma-Aldrich (35)
Physical Form
Filter Products
Showing 1-20 of 35
Page 1 of 2
Page 1 of 2
Compare
Product Number
Description
Pricing
NCXAUCR40
Gold Nanospheres,
Bare, 40 nm, for Passive Adsorption, Bioready, Bare (Citrate), OD 20, in Aqueous Sodium Citrate
Expand
View Pricing
NCXAUIR40
Gold Nanospheres,
40 nm, with Streptavidin, Bioready, OD 10, Streptavidin, in Aqueous Conjugate Buffer
Expand
View Pricing
NCXAUXR40
Gold Nanospheres,
40 nm, for Covalent Conjugation, Bioready, Carboxyl, OD 20, in Water
Expand
View Pricing
905550
Gold Nanospheres,
100 nm, FITC and carboxyl functionalized, powder
Expand
View Pricing
905933
Gold Nanospheres,
15 nm diameter, Cy3 and streptavidin functionalized, powder
Expand
View Pricing
NCXAUXU100
Gold Nanospheres,
Ultra Uniform, 100 nm, 0.05 mg/mL (Aqueous 2 mM Sodium Citrate), PEG Carboxyl 0.8 kDa
Expand
View Pricing
905755
Gold Nanospheres,
100 nm, FITC and streptavidin functionalized, powder
Expand
View Pricing
905747
Gold Nanospheres,
100 nm, FITC and NHS functionalized, powder
Expand
View Pricing
NCXAUAH100
Gold Nanospheres,
Silica-Shelled, 100 nm, NanoXact, 1 mg/mL in ethanol, Aminated / Cationic Silica
Expand
View Pricing
NCXAUSH50
Gold Nanospheres,
Silica-Shelled, 50 nm, NanoXact, 1 mg/mL in ethanol, Silica (Silanol)
Expand
View Pricing
NCXAUNR403S
Gold Nanospheres,
40 nm, for Conjugation, Bioready, Dried, NHS, Small Reaction Kit (with 3 Spin Filters)
Expand
View Pricing
905569
Gold Nanospheres,
15 nm, Cy3 and carboxyl functionalized, powder
Expand
View Pricing
NCXAUXU30
Gold Nanospheres,
Ultra Uniform, 30 nm, 0.05 mg/mL (Aqueous 2 mM Sodium Citrate), PEG Carboxyl 0.8 kDa
Expand
View Pricing
905666
Gold Nanospheres,
100 nm, FITC and maleimide functionalized, powder
Expand
View Pricing
905623
Gold Nanospheres,
15 nm, Cy3 and amine functionalized, powder
Expand
View Pricing
NCXAUXU50
Gold Nanospheres,
Ultra Uniform, 50 nm, 0.05 mg/mL (Aqueous 2 mM Sodium Citrate), PEG Carboxyl 0.8 kDa
Expand
View Pricing
NCXAUNR401L
Gold Nanospheres,
40 nm, for Conjugation, Bioready, Dried, NHS, Large Reaction Kit (with 1 Spin Filter), kit of 1 unit
Expand
View Pricing
905836
Gold Nanospheres,
100 nm, FITC and protein A functionalized, powder
Expand
View Pricing
NCXAUXR80
Gold Nanospheres,
80 nm, for Covalent Conjugation, Bioready, Carboxyl, OD 20, in Water
Expand
View Pricing
NCXAUXU10
Gold Nanospheres,
Ultra Uniform, 10 nm, 0.05 mg/mL (Aqueous 2 mM Sodium Citrate), PEG Carboxyl 0.8 kDa
Expand
View Pricing
Page 1 of 2
Page 1 of 2