Skip to Content
AI
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Gold nanorods
Synonym(s):
Au nanorods, Gold nanorod
Products
Brand
Sigma-Aldrich (33)
pH Value
Physical Form
Quality Segment
Filter Products
Showing 1-20 of 33
Page 1 of 2
Page 1 of 2
Compare
Product Number
Description
Pricing
716898
Gold nanorods,
carboxyl terminated, 10 nm diameter, λ
max
, 808 nm, dispersion in H
2
O
Expand
View Pricing
716812
Gold nanorods,
10 nm diameter, λ
max
, 780 nm, dispersion in H
2
O
Expand
View Pricing
716820
Gold nanorods,
10 nm diameter, λ
max
, 808 nm, dispersion in H
2
O
Expand
View Pricing
771686
Gold nanorods,
25 nm diameter, λ
max
, 650 nm, dispersion in H
2
O
Expand
View Pricing
900363
Gold nanorods,
10 nm diameter, λ
max
, 808 nm, dispersion in H
2
O, citrate capped
Expand
View Pricing
900364
Gold nanorods,
10 nm diameter, λ
max
, 980 nm, dispersion in H
2
O, citrate capped
Expand
View Pricing
771651
Gold nanorods,
25 nm diameter, λ
max
, 600 nm, dispersion in H
2
O
Expand
View Pricing
776688
Gold nanorods,
10 nm diameter, λ: 1064 nm A
max
, dispersion in H
2
O
Expand
View Pricing
900365
Gold nanorods,
10 nm diameter, λ
max
, 1064 nm, dispersion in H
2
O, citrate capped
Expand
View Pricing
900367
Gold nanorods,
25 nm diameter, λ
max
, 650 nm, dispersion in H
2
O, citrate capped
Expand
View Pricing
716839
Gold nanorods,
10 nm diameter, λ
max
, 850 nm, dispersion in H
2
O
Expand
View Pricing
907634
Gold nanorods,
25 nm diameter, FITC and carboxyl functionalized, powder
Expand
View Pricing
900362
Gold nanorods,
10 nm diameter, λ
max
, 780 nm, dispersion in H
2
O, citrate capped
Expand
View Pricing
776661
Gold nanorods,
10 nm diameter, λ
max
, 980 nm, dispersion in H
2
O
Expand
View Pricing
900366
Gold nanorods,
25 nm diameter, λ
max
, 550 nm, dispersion in H
2
O, citrate capped
Expand
View Pricing
716928
Gold nanorods,
palladium coated, 25 nm diameter, 100 μg/mL in H
2
O, dispersion
Expand
View Pricing
905984
Gold nanorods,
10 nm diameter, Cy3 and amine functionalized, powder
Expand
View Pricing
905895
Gold nanorods,
10 nm diameter, Cy3 and NHS functionalized, powder
Expand
View Pricing
906212
Gold nanorods,
25 nm diameter, FITC and streptavidin functionalized, powder
Expand
View Pricing
905712
Gold nanorods,
10 nm diameter, Cy3 and methyl functionalized, powder
Expand
View Pricing
Page 1 of 2
Page 1 of 2