Skip to Content
Merck
Gold(III) chloride hydrate ~52% Au basis

Gold(III) chloride hydrate

Gold(III) chloride hydrate ~52% Au basis
Synonym(s):
Auric chloride, Chloroauric acid, Hydrogen tetrachloroaurate(III) hydrate, Tetrachloroauric(III) acid
Linear Formula:
HAuCl4 · aq
CAS Number:
Molecular Weight:
339.79 (anhydrous basis)
EC Number:

Products

Brand
Color
Formula Weight
Physical Form
Purity
Chemical Composition
Quality Segment
Reagent Type
Technique
Showing 1-3 of 3