Skip to Content
AI
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Gly-Gly-Gly
Synonym(s):
Glycyl-glycyl-glycine, Triglycine
Linear Formula:
NH
2
CH
2
CONHCH
2
CONHCH
2
COOH
CAS Number:
556-33-2
Molecular Weight:
189.17
EC Number:
209-122-0
Products
Product Category
amino acids (2)
Brand
Sigma-Aldrich (1)
Color
Formula Weight
pH Value
Physical Form
Product Line
Purity
Quality Segment
Application
Technique
Filter Products
Showing 1-2 of 2
Compare
Product Number
Description
Pricing
G1377
Gly-Gly-Gly,
≥98% (TLC)
Expand
View Pricing
50239
Gly-Gly-Gly,
≥99.0% (NT), suitable for ligand binding assays, BioUltra
Expand
View Pricing