Skip to Content
AI
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Glyceryl trilinoleate
Synonym(s):
1,2,3-Tri-(cis,cis-9,12-octadecadienoyl)glycerol, 1,2,3-Trilinoleoylglycerol, Glycerol trilinoleate, Trilinolein
Empirical Formula (Hill Notation):
C
57
H
98
O
6
CAS Number:
537-40-6
Molecular Weight:
879.38
EC Number:
208-666-6
Products
Product Category
lipids (2)
glycerides (1)
metabolites (1)
Brand
Sigma-Aldrich (1)
Supelco (1)
Biological Source
Formula Weight
Physical Form
Purity
Format
Quality Segment
Functional Group
Application
Reference Material Type
Special Grade
analytical standard (1)
Technique
Filter Products
Showing 1-2 of 2
Compare
Product Number
Description
Pricing
T9517
Glyceryl trilinoleate,
≥98% (TLC), liquid
Expand
View Pricing
96373
Glyceryl trilinoleate,
analytical standard
Expand
View Pricing