Skip to Content
AI
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Water Purification
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
Get Site Smart
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Glucose-6-phosphate Dehydrogenase from baker’s yeast (
S. cerevisiae
)
Synonym(s):
G-6-P-DH, Zwischenferment
CAS Number:
9001-40-5
EC Number:
232-602-6
Products
Product Category
enzymes (1)
Brand
Sigma-Aldrich (3)
Biological Source
Physical Form
Quality Segment
Application
Technique
Filter Products
Showing 1-3 of 3
Compare
Product Number
Description
Pricing
G6378
Glucose-6-phosphate Dehydrogenase from baker’s yeast (
S. cerevisiae
),
Type XV, lyophilized powder, 200-400 units/mg protein (modified Warburg-Christian)
Expand
View Pricing
G7877
Glucose-6-phosphate Dehydrogenase from baker’s yeast (
S. cerevisiae
),
Type VII, ammonium sulfate suspension, ≥200 units/mg protein
Expand
View Pricing
G4134
Glucose-6-phosphate Dehydrogenase from baker’s yeast (
S. cerevisiae
),
Type IX, lyophilized powder, 200-400 units/mg protein (modified Warburg-Christian)
Expand
View Pricing