Skip to Content
AI
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Fractogel
®
EMD TMAE Hicap (M)
Products
Product Category
columns (4)
resins (1)
Brand
Millipore (5)
Physical Form
Product Line
Analyte Suitability
Separation Technique
ID (mm)
Volume (ml)
Quality Segment
Application
Housing Material
Particle Size (µm)
Sterility
Filter Products
Showing 1-5 of 5
Compare
Product Number
Description
Pricing
1.10316
Fractogel
®
EMD TMAE Hicap (M),
Expand
View Pricing
1.25137
Fractogel
®
EMD TMAE Hicap (M),
RoboColumn Fractogel
®
TMAE Hicap (M), volume 0.6 mL, prepacked column
Expand
View Pricing
1.25070
Fractogel
®
EMD TMAE Hicap (M),
MiniChrom Column Fractogel
®
TMAE Hicap (M), volume 5 mL, prepacked column
Expand
View Pricing
1.25085
Fractogel
®
EMD TMAE Hicap (M),
RoboColumn Fractogel
®
TMAE Hicap (M), volume 0.2 mL, prepacked column
Expand
View Pricing
1.25055
Fractogel
®
EMD TMAE Hicap (M),
MiniChrom Column Fractogel
®
TMAE Hicap (M), volume 1 mL, prepacked column
Expand
View Pricing