Skip to Content
AI
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Formamide
Synonym(s):
Amide C
1
, Formic amide, Formamide
Linear Formula:
HCONH
2
CAS Number:
75-12-7
Molecular Weight:
45.04
EC Number:
200-842-0
Products
Product Category
solvents (7)
Brand
Millipore (1)
Sigma-Aldrich (6)
Supelco (1)
Boiling Point (°C)
Color
Formula Weight
Melting Point (°C)
pH Value
Physical Form
Product Line
Purity
Research Area
Quality Segment
Application
Technique
Filter Products
Showing 1-10 of 10
Compare
Product Number
Description
Pricing
F7503
Formamide,
ReagentPlus
®
, ≥99.0% (GC)
Expand
View Pricing
F9037
Formamide,
BioReagent, ≥99.5% (GC), for molecular biology
Expand
View Pricing
47671
Formamide,
BioUltra, for molecular biology, ≥99.5% (T)
Expand
View Pricing
1.09684
Formamide,
for analysis EMSURE
®
Expand
View Pricing
4610-OP
Formamide,
OmniPur
®
Grade, >= 99%, Deionized
Expand
View Pricing
295876
Formamide,
spectrophotometric grade, ≥99%
Expand
View Pricing
1.04008
Formamide,
EMPLURA
®
Expand
View Pricing
221198
Formamide,
ACS reagent, ≥99.5%
Expand
View Pricing
11-0740
Formamide,
JIS special grade, ≥98.5%
Expand
View Pricing
11-0750
Formamide,
SAJ first grade, ≥98.5%
Expand
View Pricing