Skip to Content
AI
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Formaldehyde solution
Synonym(s):
Formalin
Linear Formula:
HCHO
CAS Number:
50-00-0
Molecular Weight:
30.03
Products
Product Category
building blocks (4)
solvents (3)
Brand
SAFC (1)
Sigma-Aldrich (8)
Supelco (1)
Feature
Formula Weight
Physical Form
Agency
Research Area
Quality Segment
Application
Special Grade
ACS reagent (1)
certified reference material (1)
Technique
Filter Products
Showing 1-10 of 10
Compare
Product Number
Description
Pricing
F8775
Formaldehyde solution,
for molecular biology, 36.5-38% in H
2
O
Expand
View Pricing
F1635
Formaldehyde solution,
contains 10-15% methanol as stabilizer, 37 wt. % in H
2
O
Expand
View Pricing
47608
Formaldehyde solution,
for molecular biology, BioReagent, ≥36.0% in H
2
O (T)
Expand
View Pricing
252549
Formaldehyde solution,
ACS reagent, 37 wt. % in H
2
O, contains 10-15% Methanol as stabilizer (to prevent polymerization)
Expand
View Pricing
47083-U
Formaldehyde solution,
stabilized with methanol, ~37 wt. % in H
2
O, certified reference material
Expand
View Pricing
15512
Formaldehyde solution,
meets analytical specification of USP, ≥34.5 wt. %
Expand
View Pricing
47673
Formaldehyde solution,
tested according to Ph. Eur.
Expand
View Pricing
11-0730
Formaldehyde solution,
SAJ first grade, ≥35.0%, contains methanol as stabilizer
Expand
View Pricing
11-0735
Formaldehyde solution,
10%
Expand
View Pricing
11-0720
Formaldehyde solution,
JIS special grade, 36.0-38.0%, contains methanol as stabilizer
Expand
View Pricing