Skip to Content
AI
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Floating tube rack
Products
Product Category
racks (4)
Brand
Sigma-Aldrich (3)
Manufacturer
Diameter (mm)
Color
Filter Products
Showing 1-4 of 4
Compare
Product Number
Description
Pricing
R7776
Floating tube rack,
rectangular shaped, holds 24 tubes, Holds 24 x 1.5-2 mL tubes
Expand
View Pricing
R7526
Floating tube rack,
diamond shaped, holds 4 tubes, Holds 4 x 50 mL tubes
Expand
View Pricing
R7651
Floating tube rack,
parallelogram shaped, holds 8 tubes, Holds 8 x 15 mL tubes
Expand
View Pricing
R7901
Floating tube rack,
circle shaped, holds 18 tubes, Holds 18 x 0.2-2 mL tubes
Expand
View Pricing