Skip to Content
Merck
FK-506 monohydrate ≥98% (HPLC)

FK-506 monohydrate

FK-506 monohydrate ≥98% (HPLC)
Synonym(s):
Tacrolimus
Empirical Formula (Hill Notation):
C44H69NO12 · H2O
CAS Number:
Molecular Weight:
822.03

Products

Product Category
Brand
Formula Weight
Gene Alias
Physical Form
Purity
Research Area
Quality Segment
Showing 1-1 of 1
Compare
Product Number
Description
Pricing