
Farnesene, mixture of isomers

Empirical Formula (Hill Notation):
C15H24
Molecular Weight:
204.35
Products
Product Category
Brand
Biological Source
Boiling Point (°C)
Color
Formula Weight
Physical Form
Agency
Chemical Composition
Quality Segment
Application
Organoleptic Class
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing