Skip to Content
Merck
Eucalyptus Oil pharmaceutical secondary standard, certified reference material

Eucalyptus oil

Eucalyptus Oil pharmaceutical secondary standard, certified reference material
Synonym(s):
Eucalyptus oil
CAS Number:

Products

Product Category
Brand
Sustainability
Feature
Manufacturer
Physical Form
Purity
Agency
Format
Quality Segment
Application
Organoleptic Class
Package Size
API Family
Reference Material Type
Special Grade