Skip to Content
AI
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Water Purification
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
Get Site Smart
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Ethylene carbonate
Synonym(s):
1,3-Dioxolan-2-one, Ethylene carbonate
Empirical Formula (Hill Notation):
C
3
H
4
O
3
CAS Number:
96-49-1
Molecular Weight:
88.06
EC Number:
202-510-0
Products
Product Category
building blocks (1)
solvents (1)
Brand
Sigma-Aldrich (3)
Supelco (1)
Boiling Point (°C)
Formula Weight
Manufacturer
Melting Point (°C)
Physical Form
Purity
Quality Segment
Application
Markush Class
Markush Group
Reference Material Type
Technique
Filter Products
Showing 1-4 of 4
Compare
Product Number
Description
Pricing
809950
Ethylene carbonate,
battery grade, ≥99%, acid <10 ppm, H2O <10 ppm
Expand
View Pricing
E26258
Ethylene carbonate,
98%
Expand
View Pricing
8.44011
Ethylene carbonate,
for synthesis
Expand
View Pricing
01380
Ethylene carbonate,
Trace
CERT
®
, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Expand
View Pricing