Skip to Content
AI
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Eppendorf
®
Upright Freezer Rack
Products
Product Category
racks (39)
Manufacturer
Diameter (mm)
Height (mm)
Technique
Filter Products
Showing 1-20 of 39
Page 1 of 2
Page 1 of 2
Compare
Product Number
Description
Pricing
EP6001021210
Eppendorf
®
Upright Freezer Rack,
Holds 16 x boxes (2 in.), (4 x 4) configuration, stainless steel, side access
Expand
View Pricing
EPK0641-3001
Eppendorf
®
Upright Freezer Rack,
Holds 20 x 76 mm/ 3 in boxes, (4 x 5) configuration, aluminum, drawer
Expand
View Pricing
EP6001062410
Eppendorf
®
Upright Freezer Rack,
Holds 12 x 102 mm/4 in boxes, (4 x 3) configuration, stainless steel, drawer
Expand
View Pricing
EP6001021910
Eppendorf
®
Upright Freezer Rack,
Holds 12 x 64 mm/2.5 in boxes, (4 x 3) configuration, stainless steel, side access
Expand
View Pricing
EP6001012310
Eppendorf
®
Upright Freezer Rack,
Holds 20 x boxes (3 in.), (4 x 5) configuration, stainless steel, drawer
Expand
View Pricing
EP6001022910
Eppendorf
®
Upright Freezer Rack,
Holds 12 x 64 mm/2.5 in boxes, (4 x 3) configuration, stainless steel, drawer
Expand
View Pricing
EP6001021410
Eppendorf
®
Upright Freezer Rack,
Holds 8 x 102 mm/4 in boxes, (4 x 2) configuration, stainless steel, side access
Expand
View Pricing
EP6001022210
Eppendorf
®
Upright Freezer Rack,
Holds 8 x 53 mm/2 in boxes, (4 x 4) configuration, stainless steel, drawer
Expand
View Pricing
EPK0641-3003
Eppendorf
®
Upright Freezer Rack,
Holds 10 x 53 mm/ 2 in boxes, (5 x 2) configuration, aluminum, drawer
Expand
View Pricing
EPK0641-3002
Eppendorf
®
Upright Freezer Rack,
Holds 12 x 102 mm/ 4 in boxes, (4 x 3) configuration, aluminum, drawer
Expand
View Pricing
EPK0641-1900
Eppendorf
®
Upright Freezer Rack,
Holds 8 x 53 mm/ 2 in boxes, (4 x 4) configuration, aluminum, drawer
Expand
View Pricing
EPK0641-3004
Eppendorf
®
Upright Freezer Rack,
Holds 6 x 76 mm/ 3 in boxes, (3 x 2) configuration, aluminum, drawer
Expand
View Pricing
EP6001012210
Eppendorf
®
Upright Freezer Rack,
Holds 28 x boxes (2 in.), (4 x 7) configuration, stainless steel, drawer
Expand
View Pricing
EP6001012910
Eppendorf
®
Upright Freezer Rack,
Holds 24 x boxes (2.5 in.), (4 x 6) configuration, stainless steel, drawer
Expand
View Pricing
EPK0641-3000
Eppendorf
®
Upright Freezer Rack,
Holds 28 x 53 mm/ 2 in boxes, (4 x 7) configuration, aluminum, drawer
Expand
View Pricing
EP6001062210
Eppendorf
®
Upright Freezer Rack,
Holds 24 x 53 mm/2 in boxes, (4 x 6) configuration, stainless steel, drawer
Expand
View Pricing
EP6001062510
Eppendorf
®
Upright Freezer Rack,
Holds 8 x 127 mm/5 in boxes, (4 x 2) configuration, stainless steel, drawer
Expand
View Pricing
EPK0641-1890
Eppendorf
®
Upright Freezer Rack,
Holds 8 x 76 mm/ 3 in boxes, (4 x 4) configuration, aluminum, drawer
Expand
View Pricing
EP6001012410
Eppendorf
®
Upright Freezer Rack,
Holds 12 x boxes (4 in.), (4 x 3) configuration, stainless steel, drawer
Expand
View Pricing
EP6001021110
Eppendorf
®
Upright Freezer Rack,
Holds 24 x DWP (4 x 6) configuration, stainless steel, side access
Expand
View Pricing
Page 1 of 2
Page 1 of 2