Skip to Content
AI
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Water Purification
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
Get Site Smart
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Eppendorf Tubes
®
BioBased
Synonym(s):
sustainable single-use plastic
Products
Product Category
tubes (4)
Sustainability
Color
Feature
Manufacturer
Capacity (mL)
Color
Package Size
Sterility
Filter Products
Showing 1-4 of 4
Compare
Product Number
Description
Pricing
EP0030122518
Eppendorf Tubes
®
BioBased,
capacity 5.0 mL, sterile, screw top cap, pack of 2 × 100 ea
Expand
View Pricing
EP0030122542
Eppendorf Tubes
®
BioBased,
capacity 50 mL, sterile, screw top cap, pack of 20 × 25 ea
Expand
View Pricing
EP0030122526
Eppendorf Tubes
®
BioBased,
capacity 15 mL, sterile, screw top cap, pack of 10 × 50 ea
Expand
View Pricing
EP0030122534
Eppendorf Tubes
®
BioBased,
capacity 25 mL, sterile, screw top cap, pack of 8 × 25 ea
Expand
View Pricing