Skip to Content
AI
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Eppendorf Tube
®
Products
Product Category
tubes (6)
Color
Manufacturer
Capacity (mL)
Volume (ml)
Color
Package Size
Sterility
Technique
Filter Products
Showing 1-6 of 6
Compare
Product Number
Description
Pricing
EP0030119401
Eppendorf Tube
®
,
capacity 5.0 mL, Eppendorf Quality
™
, pack of 200 ea (2 bags of 100 ea)
Expand
View Pricing
EP0030119487
Eppendorf Tube
®
,
capacity 5.0 mL, sterile, pack of 200 pieces (10 x 20 ea)
Expand
View Pricing
EP0030119460
Eppendorf Tube
®
,
capacity 5.0 mL, PCR clean, pack of 200 pieces (2 bags x 100 ea)
Expand
View Pricing
EP0030119479
Eppendorf Tube
®
,
capacity 5.0 mL, Biopur
®
, pack of 50 ea (individuallt wrapped)
Expand
View Pricing
EP0030122330
Eppendorf Tube
®
,
amber tube, tube volume 5 mL, PCR clean, light protection, case of 200 ea (2 bags of 100 Tubes each)
Expand
View Pricing
EP0030119380
Eppendorf Tube
®
,
capacity 5.0 mL (5.0), PCR clean, Starter Pack, PCR clean, 400 Tubes, 2 Tube-Racks, 8 adapters for rotors with bore for 15 mL conical tubes
Expand
View Pricing