Skip to Content
AI
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Eppendorf
®
IsoTherm-System
®
Products
Product Category
racks (3)
Manufacturer
Package Size
Filter Products
Showing 1-4 of 4
Compare
Product Number
Description
Pricing
ISOTS5-EP
Eppendorf
®
IsoTherm-System
®
,
IsoPack and IsoRack set, includes one IsoPack and one IsoRack, for 0.5 mL vessels, -21 °C
Expand
View Pricing
ISOTSS2-EP
Eppendorf
®
IsoTherm-System
®
,
starter set, 0 °C IsoPack and -21 °C IsoPack, for tubes, 1.5/2.0 mL, Eppendorf
®
022510053(NA)
Expand
View Pricing
ISOTS4R-EP
Eppendorf
®
IsoTherm-System
®
,
IsoRack, set of 4 racks, for 1.5 mL/2.0 mL vessels
Expand
View Pricing
ISOTSS5-EP
Eppendorf
®
IsoTherm-System
®
,
starter set, includes IsoSafe, IsoRack, 0 °C IsoPack and -21 °C IsoPack, for tubes, 0.5 mL
Expand
View Pricing