Skip to Content
Products
Cart
0
AI
EN
Products
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Water Purification
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
Get Site Smart
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Login
Back
Eppendorf
®
Flex-Tubes Microcentrifuge Tubes
Products
Product Category
tubes (7)
vials (1)
Manufacturer
Volume (ml)
Color
Package Size
Sterility
Technique
Filter Products
Showing 1-7 of 7
Compare
Product Number
Description
Pricing
EP022364120
Eppendorf
®
Flex-Tubes Microcentrifuge Tubes,
volume 1.5 mL, colorless, suitable for PCR, pack of 500 tubes
Expand
View Pricing
EP022363531
Eppendorf
®
Flex-Tubes Microcentrifuge Tubes,
volume 1.5 mL, blue, pack of 500 tubes
Expand
View Pricing
EP022364111
Eppendorf
®
Flex-Tubes Microcentrifuge Tubes,
clear, capacity 1.5 mL, pack of 500 ea
Expand
View Pricing
EP022363514
Eppendorf
®
Flex-Tubes Microcentrifuge Tubes,
volume 1.5 mL, amber, pack of 500 tubes
Expand
View Pricing
EP022363590
Eppendorf
®
Flex-Tubes Microcentrifuge Tubes,
volume 1.5 mL, yellow, pack of 500 tubes
Expand
View Pricing
EP022363573
Eppendorf
®
Flex-Tubes Microcentrifuge Tubes,
volume 1.5 mL, red, pack of 500 tubes
Expand
View Pricing
EP022363557
Eppendorf
®
Flex-Tubes Microcentrifuge Tubes,
volume 1.5 mL, green, pack of 500 tubes
Expand
View Pricing
Choose up to 4 products to compare
Compare
Clear All
English - EN
Learn More