Skip to Content
AI
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Eppendorf
®
Chest Freezer Rack
Products
Product Category
racks (22)
Manufacturer
Diameter (mm)
Height (mm)
Technique
Filter Products
Showing 1-20 of 22
Page 1 of 2
Page 1 of 2
Compare
Product Number
Description
Pricing
EP6001050211
Eppendorf
®
Chest Freezer Rack,
Holds 13 x 53 mm/2 in boxes, (13 x 1) configuration, stainless steel, side access, with locking rod
Expand
View Pricing
EP6001040211
Eppendorf
®
Chest Freezer Rack,
Holds 14 x 53 mm/2 in boxes, (14 x 1) configuration, stainless steel, side access, with locking rod
Expand
View Pricing
EP6001000911
Eppendorf
®
Chest Freezer Rack,
Holds 11 x boxes (2.5 in.), (11 x 1) configuration, stainless steel, side access, with locking rod
Expand
View Pricing
EPK0641-1750
Eppendorf
®
Chest Freezer Rack,
Holds 6 x 102 mm/ 4 in boxes, (1 x 6) configuration, aluminum, side access
Expand
View Pricing
EP6001040210
Eppendorf
®
Chest Freezer Rack,
Holds 14 x 53 mm/2 in boxes, (1 x 14) configuration, stainless steel, side access
Expand
View Pricing
EP6001000311
Eppendorf
®
Chest Freezer Rack,
Holds 9 x boxes (3 in.), (9 x 1) configuration, stainless steel, side access, with locking rod
Expand
View Pricing
EP6001040110
Eppendorf
®
Chest Freezer Rack,
Holds 14 x DWP (53 mm/2 in), (14 x 1) configuration, stainless steel, side access
Expand
View Pricing
EP6001050210
Eppendorf
®
Chest Freezer Rack,
Holds 13 x 53 mm/2 in boxes, (13 x 1) configuration, stainless steel, side access
Expand
View Pricing
EP6001000310
Eppendorf
®
Chest Freezer Rack,
Holds 9 x boxes (3 in.), (9 x 1) configuration, stainless steel, side access
Expand
View Pricing
EP6001040111
Eppendorf
®
Chest Freezer Rack,
Holds 14 x DWP (53 mm/ 2 in), (14 x 1) configuration, stainless steel, side access, with locking rod
Expand
View Pricing
EP6001000510
Eppendorf
®
Chest Freezer Rack,
Holds 5 x boxes (5 in.), (5 x 1) configuration, stainless steel, side access
Expand
View Pricing
EP6001040011
Eppendorf
®
Chest Freezer Rack,
Holds 14 x MTP (18 mm), (14 x 1) configuration, stainless steel, side access, with locking rod
Expand
View Pricing
EP6001050010
Eppendorf
®
Chest Freezer Rack,
Holds 37 x MTP (18 mm), (37 x 1) configuration, stainless steel, side access
Expand
View Pricing
EP6001000411
Eppendorf
®
Chest Freezer Rack,
Holds 7 x boxes (4 in.), (7 x 1) configuration, stainless steel, side access, with locking rod
Expand
View Pricing
EP6001050011
Eppendorf
®
Chest Freezer Rack,
Holds 37 x MTP (18 mm), (37 x 1) configuration, stainless steel, side access, with locking rod
Expand
View Pricing
EP6001000910
Eppendorf
®
Chest Freezer Rack,
Holds 11 x boxes (2.5 in.), (11 x 1) configuration, stainless steel, side access
Expand
View Pricing
EP6001050110
Eppendorf
®
Chest Freezer Rack,
Holds 13 x DWP (53 mm), (13 x 1) configuration, stainless steel, side access
Expand
View Pricing
EP6001050111
Eppendorf
®
Chest Freezer Rack,
Holds 13 x DWP (53 mm), (13 x 1) configuration, stainless steel, side access, with locking rod
Expand
View Pricing
EP6001040010
Eppendorf
®
Chest Freezer Rack,
Holds 38 x MTP (18 mm), (38 x 1) configuration, stainless steel, side access
Expand
View Pricing
EP6001000410
Eppendorf
®
Chest Freezer Rack,
Holds 7 x boxes (4 in.), (7 x 1) configuration, stainless steel, side access
Expand
View Pricing
Page 1 of 2
Page 1 of 2