Skip to Content
AI
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Eppendorf
®
Centrifuge 5910 Ri G
Synonym(s):
Eppendorf
®
Centrifuge 5910 Ri G
Products
Product Category
centrifuges (6)
Manufacturer
Package Size
Technique
Filter Products
Showing 1-6 of 6
Compare
Product Number
Description
Pricing
EP5943000106
Eppendorf
®
Centrifuge 5910 Ri G,
refrigerated, with Rotor FA-6x50, AC/DC input 120 V, 50 - 60 Hz, US 3-pin plug
Expand
View Pricing
EP5943000548
Eppendorf
®
Centrifuge 5910 Ri G,
refrigerated, with S-4x750 rotor, AC/DC input 120 V, 50 - 60 Hz, US 3-pin plug
Expand
View Pricing
EP5943000343
Eppendorf
®
Centrifuge 5910 Ri G,
refrigerated, with Rotor S-4xUniversal, incl. universal buckets and adapters for 5 mL/15 mL/50 mLconical tubes and plates, AC/DC input 120 V, 50 - 60 Hz, US 3-pin plug
Expand
View Pricing
EP5943000246
Eppendorf
®
Centrifuge 5910 Ri G,
refrigerated, with Rotor S-4xUniversal, incl. universal buckets and adapters for 5 mL/15 mL/50 mLconical tubes, 175 – 250 mL bottles and plates, AC/DC input 120 V, 50 - 60 Hz, US 3-pin plug
Expand
View Pricing
EP5943000114
Eppendorf
®
Centrifuge 5910 Ri G,
refrigerated, with Rotor S-4xUniversal, incl. universal buckets and 1 L adapters (4x), Eppendorf 1 L bottles (4x), AC/DC input 120 V, 50 - 60 Hz, US 3-pin plug
Expand
View Pricing
EP5943000131
Eppendorf
®
Centrifuge 5910 Ri G,
refrigerated, without rotor, AC/DC input 120 V, 50 - 60 Hz, US 3-pin plug
Expand
View Pricing