
Eosin Y (yellowish) (C.I. 45380)

Synonym(s):
Eosin Y (yellowish) (C.I. 45380)
Empirical Formula (Hill Notation):
C20H6Br4Na2O5
CAS Number:
Molecular Weight:
691.85
Products
Brand
Formula Weight
Physical Form
Chemical Composition
Quality Segment
Application
Showing 1-2 of 2
Compare
Product Number
Description
Pricing