Skip to Content
Merck
EDDP perchlorate solution 1.0 mg/mL in methanol (as pyrolinium), ampule of 1 mL, certified reference material, Cerilliant®

EDDP perchlorate solution

EDDP perchlorate solution 1.0 mg/mL in methanol (as pyrolinium), ampule of 1 mL, certified reference material, Cerilliant®
Empirical Formula (Hill Notation):
C20H24ClNO4
CAS Number:
Molecular Weight:
377.86
EC Number:

Products

Brand
Formula Weight
Manufacturer
Physical Form
Format
Quality Segment
Application
Package Size
Reference Material Type
Special Grade
Technique