Skip to Content
AI
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Dural
®
- aluminum/copper/magnesium
Products
Manufacturer
Physical Form
Length (mm)
OD (mm)
Filter Products
Showing 1-4 of 4
Compare
Product Number
Description
Pricing
GF40322281
Dural
®
- aluminum/copper/magnesium,
tube, Al 95%/Cu 4%/Mg 1%, outside diameter 0.71 mm, length 100 mm, wall thickness 0.051 mm
Expand
View Pricing
GF66883304
Dural
®
- aluminum/copper/magnesium,
tube, Al95%/Cu 4%/Mg 1%, outside diameter 0.71 mm, length 1000 mm, wall thickness 0.051 mm
Expand
View Pricing
GF40285885
Dural
®
- aluminum/copper/magnesium,
tube, Al 95%/Cu 4%/Mg 1%, outside diameter 0.71 mm, length 200 mm, wall thickness 0.051 mm
Expand
View Pricing
GF35735069
Dural
®
- aluminum/copper/magnesium,
tube, Al 95%/Cu 4%/Mg 1%, outside diameter 0.71 mm, length 500 mm, wall thickness 0.051 mm
Expand
View Pricing