Skip to Content
AI
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
DL-Isoleucine
Synonym(s):
DL-2-Amino-3-methylpentanoic acid
Linear Formula:
CH
3
CH
2
CH(CH
3
)CH(NH
2
)COOH
CAS Number:
443-79-8
Molecular Weight:
131.17
EC Number:
207-139-8
Products
Brand
Sigma-Aldrich (1)
Formula Weight
Melting Point (°C)
Product Line
Purity
Application
Markush Class
Markush Group
Optical Isomer
Amino Acid
Reaction Type
Filter Products
Showing 1-1 of 1
Compare
Product Number
Description
Pricing
298689
DL-Isoleucine,
ReagentPlus
®
, 99%
Expand
View Pricing