Skip to Content
AI
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
(±)-Baclofen
Synonym(s):
(±)-β-(Aminomethyl)-4-chlorobenzenepropanoic acid, Lioresal
Empirical Formula (Hill Notation):
C
10
H
12
ClNO
2
CAS Number:
1134-47-0
Molecular Weight:
213.66
EC Number:
214-486-9
Products
Brand
Sigma-Aldrich (1)
Color
Formula Weight
Gene Alias
Physical Form
Purity
Filter Products
Showing 1-1 of 1
Compare
Product Number
Description
Pricing
B5399
(±)-Baclofen,
≥98% (HPLC), solid
Expand
View Pricing