Skip to Content
AI
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
Get Site Smart
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Dimethylglyoxime
Synonym(s):
Dimethylglyoxime, 2,3-Butanedione dioxime, Diacetyldioxime
Linear Formula:
CH
3
C(=NOH)C(=NOH)CH
3
CAS Number:
95-45-4
Molecular Weight:
116.12
EC Number:
202-420-1
Products
Product Category
building blocks (3)
Brand
Sigma-Aldrich (4)
Supelco (1)
Formula Weight
Melting Point (°C)
Physical Form
Purity
Agency
Quality Segment
Functional Group
Markush Class
Markush Group
Special Grade
ACS reagent (2)
Puriss (1)
Puriss P.A. (1)
Technique
Filter Products
Showing 1-5 of 5
Compare
Product Number
Description
Pricing
1.03062
Dimethylglyoxime,
GR for analysis (reagent for nickel) ACS,Reag. Ph Eur
Expand
View Pricing
162574
Dimethylglyoxime,
ACS reagent, ≥99%
Expand
View Pricing
03858
Dimethylglyoxime,
≥97.0% (TLC)
Expand
View Pricing
40390
Dimethylglyoxime,
puriss. p.a., ACS reagent, for the detection of Ni, ≥99.0% (TLC)
Expand
View Pricing
07-4480
Dimethylglyoxime,
JIS special grade, ≥98.0%
Expand
View Pricing