Skip to Content
Merck
Dichlorotriphenylphosphorane 95%

Dichlorotriphenylphosphorane

Dichlorotriphenylphosphorane 95%
Synonym(s):
Triphenylphosphine dichloride
Linear Formula:
(C6H5)3PCl2
CAS Number:
Molecular Weight:
333.19

Products

Product Category
Formula Weight
Melting Point (°C)
Physical Form
Purity
Functional Group
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing