
Diallyl N,N-diisopropylphosphoramidite

Synonym(s):
N,N-Bis(1-methylethyl)phosphoramidous acid di-2-propenyl ester, Diallyloxy(diisopropylamino)phosphine
Linear Formula:
[(CH3)2CH]2NP(OCH2CH=CH2)2
CAS Number:
Molecular Weight:
245.30
Products
Product Category
Brand
Boiling Point (°C)
Formula Weight
Physical Form
Functional Group
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing