Skip to Content
AI
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Diaion
®
HP-20
CAS Number:
9052-95-3
Products
Product Category
resins (2)
Brand
Sigma-Aldrich (1)
Supelco (2)
Physical Form
Separation Technique
Particle Size (µm)
Pore Size (Å)
Technique
Filter Products
Showing 1-3 of 3
Compare
Product Number
Description
Pricing
13606
Diaion
®
HP-20,
pkg of 500 g
Expand
View Pricing
13607
Diaion
®
HP-20,
pkg of 1000 g
Expand
View Pricing
13605
Diaion
®
HP-20,
pkg of 100 g
Expand
View Pricing