Skip to Content
AI
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Density Standard
EC Number:
289-220-8
Products
Manufacturer
Agency
Format
Quality Segment
Application
Reference Material Type
Special Grade
certified reference material (6)
Filter Products
Showing 1-6 of 6
Compare
Product Number
Description
Pricing
CRMDEGO
Density Standard,
ASTM D 4052; ISO 12185 (0.83294g/mL, 15 °C)
Expand
View Pricing
CRMUDEKR
Density Standard,
ASTM D4052; ISO 12185 (0.79672 g/mL, 15 °C)
Expand
View Pricing
CRMUDELU
Density Standard,
ASTM D 4052; ISO 12185 (0.8687 g/mL, 15 °C)
Expand
View Pricing
CRMDEGA
Density Standard,
ASTM D 4052; ISO 12185 (0.73576 g/mL, 15 °C)
Expand
View Pricing
CRMUDEGO
Density Standard,
ASTM D 4052; IP 365 / ISO 12185 (0.8355g/mL, 15 °C)
Expand
View Pricing
CRMUDEGA
Density Standard,
ASTM D 4052; IP 365 / ISO 12185 (0.7377g/mL, 15 °C)
Expand
View Pricing