Skip to Content
AI
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Cucurbitacin E-2-O-glucoside
Synonym(s):
Elaterinide
Empirical Formula (Hill Notation):
C
38
H
54
O
13
CAS Number:
1398-78-3
Molecular Weight:
718.83
EC Number:
215-745-9
Products
Formula Weight
Manufacturer
Product Line
Format
Application
Reference Material Type
Special Grade
analytical standard (1)
Filter Products
Showing 1-1 of 1
Compare
Product Number
Description
Pricing
PHL89613
Cucurbitacin E-2-O-glucoside,
phyproof
®
Reference Substance
Expand
View Pricing