Skip to Content
AI
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Corning
®
universal fit hinged racked pipet tips
Products
Product Category
pipette tips (6)
Manufacturer
Volume Range (µL)
Color
Package Size
Sterility
Filter Products
Showing 1-6 of 6
Compare
Product Number
Description
Pricing
CLS4714
Corning
®
universal fit hinged racked pipet tips,
100-1000 μL pipet tips, blue, sterile, 100tips/rack, 10 racks/case, 1000 tips/case
Expand
View Pricing
CLS4711
Corning
®
universal fit hinged racked pipet tips,
1-200 μL pipet tips, yellow, sterile, 10 racks/case, 960 tips/case
Expand
View Pricing
CLS4710
Corning
®
universal fit hinged racked pipet tips,
1-200 μL pipet tips, yellow, non-sterile, 96 tips/rack, 10 racks/case, 960 tips/case
Expand
View Pricing
CLS4713
Corning
®
universal fit hinged racked pipet tips,
100-1000 μL pipet tips, blue, non-sterile, 10 racks/case, 1000 tips/case
Expand
View Pricing
CLS4715
Corning
®
universal fit hinged racked pipet tips,
pipet tip hinged rack insert, 100-1000 μL pipet tips, blue, non-sterile, 100 tips/rack, 10 racks/case, 1000 tips/case
Expand
View Pricing
CLS4712
Corning
®
universal fit hinged racked pipet tips,
1-200 μL pipet tips, yellow, non-sterile, 10 racks/case, 960 tips/case
Expand
View Pricing