Skip to Content
AI
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Corning
®
Transwell
®
7 well plates
Products
Product Category
plates (1)
Color
Manufacturer
Binding Type
Diameter (mm)
Surface Area cm²
Wells
Working Volume (ml)
Color
Membrane Material
Plate Material
Package Size
Pore size (µm)
Sterility
Filter Products
Showing 1-1 of 1
Compare
Product Number
Description
Pricing
CLS3452
Corning
®
Transwell
®
7 well plates,
3.0 μm pore size, Tissue Culture (TC)-treated surface clear polystyrene plate, round wells, polyester membrane (Polyethylene Terephthalate (PET), flat bottom wells clear sterile, pack of 6, case of 24
Expand
View Pricing