Skip to Content
AI
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Corning
®
organic chemistry glassware kit
Synonym(s):
Corning
®
6949E and 6949K
Products
Manufacturer
Filter Products
Showing 1-4 of 4
Compare
Product Number
Description
Pricing
CLS6949EIO
Corning
®
organic chemistry glassware kit,
foam insert only
Expand
View Pricing
CLS6949K
Corning
®
organic chemistry glassware kit,
with 14/20 joints
Expand
View Pricing
CLS6949E
Corning
®
organic chemistry glassware kit,
with 19/22 joints
Expand
View Pricing
CLS6949EBO
Corning
®
organic chemistry glassware kit,
polyethylene kit box only for 19/22 jointed kit
Expand
View Pricing