Skip to Content
AI
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Water Purification
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
Get Site Smart
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Cobalt(II) chloride hexahydrate
Synonym(s):
Cobaltous chloride hexahydrate, Cobalt(II) chloride hexahydrate
Linear Formula:
CoCl
2
· 6H
2
O
CAS Number:
7791-13-1
Molecular Weight:
237.93
EC Number:
231-589-4
Products
Product Category
salts (6)
Brand
Sigma-Aldrich (4)
Supelco (3)
Sustainability
Feature
Formula Weight
Physical Form
Product Line
Purity
Agency
Quality Segment
Reagent Type
Core
Special Grade
ACS reagent (2)
Puriss (1)
Puriss P.A. (1)
Technique
Filter Products
Showing 1-12 of 12
Compare
Product Number
Description
Pricing
255599
Cobalt(II) chloride hexahydrate,
ACS reagent, 98%
Expand
View Pricing
202185
Cobalt(II) chloride hexahydrate,
reagent grade
Expand
View Pricing
C8661
Cobalt(II) chloride hexahydrate,
BioReagent, suitable for cell culture, suitable for insect cell culture
Expand
View Pricing
31277
Cobalt(II) chloride hexahydrate,
puriss. p.a., ACS reagent, reag. Ph. Eur., 98-102%
Expand
View Pricing
1.02539
Cobalt(II) chloride hexahydrate,
for analysis EMSURE
®
ACS,Reag. Ph Eur
Expand
View Pricing
769495
Cobalt(II) chloride hexahydrate,
≥97%
Expand
View Pricing
C2911
Cobalt(II) chloride hexahydrate,
suitable for plant cell culture
Expand
View Pricing
1.02533
Cobalt(II) chloride hexahydrate,
extra pure
Expand
View Pricing
1.93652
Cobalt(II) chloride hexahydrate,
EMPLURA
®
Expand
View Pricing
935743
Cobalt(II) chloride hexahydrate,
≥99.99% trace metals basis
Expand
View Pricing
05-5150
Cobalt(II) chloride hexahydrate,
JIS special grade, ≥99.0%
Expand
View Pricing
05-5160
Cobalt(II) chloride hexahydrate,
SAJ first grade, ≥95.0%
Expand
View Pricing