
Clothianidin

Synonym(s):
(E)-1-(2-Chloro-5-thiazolylmethyl)-3-methyl-2-nitroguanidine
Empirical Formula (Hill Notation):
C6H8ClN5O2S
CAS Number:
Molecular Weight:
249.68
Products
Product Category
Brand
Formula Weight
Manufacturer
Product Line
Format
Quality Segment
Application
Reference Material Type
Special Grade
Technique
Showing 1-2 of 2