Skip to Content
AI
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Clinical 4-Way Tube Rack
Products
Product Category
racks (5)
Manufacturer
Height (mm)
Length (mm)
Color
Package Size
Filter Products
Showing 1-5 of 5
Compare
Product Number
Description
Pricing
HS120683
Clinical 4-Way Tube Rack,
assorted colors
Expand
View Pricing
HS120687
Clinical 4-Way Tube Rack,
clear
Expand
View Pricing
HS120685
Clinical 4-Way Tube Rack,
purple
Expand
View Pricing
HS120684
Clinical 4-Way Tube Rack,
blue
Expand
View Pricing
HS120686
Clinical 4-Way Tube Rack,
green
Expand
View Pricing